siRNA Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012979.1 | - | 1 | 1143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV570520.1 | - | 1 | 146 | Chr. 4 - 100443432 - 100443957 [+] on Build GRCm38 | |

Ror1
Genomic Map