siRNA Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012979.1 | - | 1 | 1588 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI102835.1 | - | 1 | 446 | Chr. 4 - 100443571 - 100444243 [+] on Build GRCm38 | |

Ror1
Genomic Map