siRNA Details
| Gene Symbol: 1700023G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006276.1 | - | 1 | 351 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706357.1 | - | 1 | 351 | Chr. 7 - 134261315 - 134261882 [-] on Build GRCm38 | |

1700023G08Rik
Genomic Map