siRNA Details
| Gene Symbol: Mib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048082.1 | - | 1 | 2489 | ||
| AK080083.1 | - | 1 | 405 | |||
| AK080847.1 | - | 15 | 3122 | |||
| AK088539.1 | - | 1 | 2794 | |||
| AK161848.1 | - | 7 | 1868 | |||
| BC083072.1 | - | 11 | 6032 | |||

Mib1
Genomic Map