siRNA Details
| Gene Symbol: Gm15743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036265.1 | - | 1 | 1512 | ||
| BB131186.2 | - | 1 | 301 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY538086.1 | - | 1 | 44 | Chr. 16 - 31427879 - 31428309 [-] on Build GRCm38 | |

Gm15743
Genomic Map