siRNA Details
| Gene Symbol: A530006G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046014.1 | 3 | 1051 | |||
| GenBank mRNA | AK079921.1 | - | 3 | 1052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A530006G24Rik| Gene Symbol: A530006G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046014.1 | 3 | 1051 | |||
| GenBank mRNA | AK079921.1 | - | 3 | 1052 | ||