siRNA Details
| Gene Symbol: C030005K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021048.1 | - | 4 | 438 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728108.1 | - | 4 | 435 | Chr. 18 - 50042730 - 50053660 [-] on Build GRCm38 | |

C030005K06Rik
Genomic Map