siRNA Details
| Gene Symbol: 5730420D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045338.1 | 3 | 526 | |||
| NR_045339.1 | 3 | 502 | ||||
| GenBank mRNA | AK017586.1 | - | 3 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730420D15Rik| Gene Symbol: 5730420D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045338.1 | 3 | 526 | |||
| NR_045339.1 | 3 | 502 | ||||
| GenBank mRNA | AK017586.1 | - | 3 | 527 | ||