siRNA Details
| Gene Symbol: 5730420D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045338.1 | 4 | 639 | |||
| NR_045339.1 | 4 | 615 | ||||
| GenBank mRNA | AK017586.1 | - | 4 | 640 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717364.1 | - | 4 | 634 | Chr. 10 - 95417380 - 95428250 [+] on Build GRCm38 | |

5730420D15Rik
Genomic Map