siRNA Details
| Gene Symbol: A930037G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020939.1 | - | 1 | 458 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742361.1 | - | 1 | 458 | Chr. 5 - 22781340 - 22782009 [+] on Build GRCm38 | |

A930037G07Rik
Genomic Map