siRNA Details
| Gene Symbol: 1700061H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006853.1 | - | 1 | 90 | ||
| AK076657.1 | - | 1 | 77 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706839.1 | - | 1 | 92 | Chr. 18 - 69943318 - 69969609 [-] on Build GRCm38 | |

1700061H18Rik
Genomic Map