siRNA Details
Gene Symbol: 1700061F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038180.1 | 4 | 852 | |||
GenBank mRNA | AK006852.1 | - | 4 | 853 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700061F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038180.1 | 4 | 852 | |||
GenBank mRNA | AK006852.1 | - | 4 | 853 |