siRNA Details
| Gene Symbol: 6030443J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102315.1 | 3 | 2487 | |||
| GenBank mRNA | AK029857.1 | - | 3 | 2464 | ||
| AK031505.1 | - | 1 | 2420 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030443J06Rik| Gene Symbol: 6030443J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102315.1 | 3 | 2487 | |||
| GenBank mRNA | AK029857.1 | - | 3 | 2464 | ||
| AK031505.1 | - | 1 | 2420 | |||