siRNA Details
| Gene Symbol: Fam222b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041269.1 | - | 1 | 234 | ||
| AK079318.1 | - | 2 | 3604 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB226091.2 | - | 1 | 273 | Chr. 11 - 78097912 - 78098397 [+] on Build GRCm38 | |

Fam222b
Genomic Map