siRNA Details
| Gene Symbol: Fam222b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041269.1 | - | 1 | 16 | ||
| AK079318.1 | - | 2 | 3386 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637765.1 | - | 1 | 16 | Chr. 11 - 78097913 - 78098409 [+] on Build GRCm38 | |

Fam222b
Genomic Map