siRNA Details
| Gene Symbol: Gm16159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110510.1 | 1 | 13 | |||
| GenBank mRNA | AK133017.1 | - | 1 | 14 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615746.1 | - | 1 | 12 | Chr. 8 - 25553515 - 25556082 [-] on Build GRCm38 | |

Gm16159
Genomic Map