siRNA Details
| Gene Symbol: Gm16159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110510.1 | 5 | 836 | |||
| GenBank mRNA | AK133017.1 | - | 5 | 838 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB015528.2 | - | 3 | 523 | Chr. 8 - 25548671 - 25554575 [-] on Build GRCm38 | |

Gm16159
Genomic Map