siRNA Details
| Gene Symbol: Gm19272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133014.1 | - | 3 | 661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19272| Gene Symbol: Gm19272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133014.1 | - | 3 | 661 | ||