siRNA Details
| Gene Symbol: 1700042G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038178.1 | 3 | 1953 | |||
| NR_038179.1 | 3 | 1906 | ||||
| GenBank mRNA | AK133010.1 | - | 3 | 1908 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV270740.2 | - | 1 | 525 | Chr. 4 - 57359797 - 57360392 [-] on Build GRCm38 | |

1700042G15Rik
Genomic Map