siRNA Details
| Gene Symbol: 1700042G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038178.1 | 3 | 1266 | |||
| NR_038179.1 | 3 | 1219 | ||||
| GenBank mRNA | AK133010.1 | - | 3 | 1220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700042G15Rik| Gene Symbol: 1700042G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038178.1 | 3 | 1266 | |||
| NR_038179.1 | 3 | 1219 | ||||
| GenBank mRNA | AK133010.1 | - | 3 | 1220 | ||