siRNA Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086423.1 | - | 1 | 644 | ||
| AK142756.1 | - | 1 | 644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxfp1| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086423.1 | - | 1 | 644 | ||
| AK142756.1 | - | 1 | 644 | |||