siRNA Details
| Gene Symbol: Rn45s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046233.2 | 1449 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rn45s| Gene Symbol: Rn45s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046233.2 | 1449 | ||||