siRNA Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082097.1 | - | 1 | 626 | ||
| AK155090.1 | - | 1 | 909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kansl1l| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082097.1 | - | 1 | 626 | ||
| AK155090.1 | - | 1 | 909 | |||