siRNA Details
| Gene Symbol: 2010015M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008259.1 | - | 1 | 633 | ||
| AK041907.1 | - | 1 | 432 | |||
| AV065408.1 | - | 1 | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010015M23Rik| Gene Symbol: 2010015M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008259.1 | - | 1 | 633 | ||
| AK041907.1 | - | 1 | 432 | |||
| AV065408.1 | - | 1 | 150 | |||