siRNA Details
Gene Symbol: C630044B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047732.1 | - | 1 | 2153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB357034.2 | - | 1 | 624 | Chr. 13 - 58813682 - 58814363 [+] on Build GRCm38 |
