siRNA Details
| Gene Symbol: C87977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139502.1 | - | 4 | 1589 | ||
| BC094895.1 | - | 4 | 1561 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C87977| Gene Symbol: C87977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139502.1 | - | 4 | 1589 | ||
| BC094895.1 | - | 4 | 1561 | |||