siRNA Details
| Gene Symbol: C87977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139502.1 | - | 4 | 1250 | ||
| BC094895.1 | - | 4 | 1223 | |||
| Gene Symbol: Oog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB077322.2 | - | 1 | 1496 | ||

C87977
Genomic Map