siRNA Details
| Gene Symbol: 4933404K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046038.1 | 4 | 1337 | |||
| GenBank mRNA | AK016652.1 | - | 4 | 1330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933404K08Rik| Gene Symbol: 4933404K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046038.1 | 4 | 1337 | |||
| GenBank mRNA | AK016652.1 | - | 4 | 1330 | ||