siRNA Details
| Gene Symbol: A330072L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039611.1 | - | 4 | 857 | ||
| AK079662.1 | - | 4 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330072L02Rik| Gene Symbol: A330072L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039611.1 | - | 4 | 857 | ||
| AK079662.1 | - | 4 | 857 | |||