siRNA Details
| Gene Symbol: Gm33335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089049.1 | - | 2 | 1028 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY763517.1 | - | 2 | 505 | Chr. 18 - 50013635 - 50016315 [-] on Build GRCm38 | |

Gm33335
Genomic Map