siRNA Details
| Gene Symbol: Gm33335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089049.1 | - | 1 | 356 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY747455.1 | - | 1 | 356 | Chr. 18 - 50014182 - 50016837 [-] on Build GRCm38 | |

Gm33335
Genomic Map