siRNA Details
Gene Symbol: Gm33335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089049.1 | - | 1 | 105 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY747455.1 | - | 1 | 105 | Chr. 18 - 50014182 - 50016837 [-] on Build GRCm38 |
