siRNA Details
Gene Symbol: Gm33335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089049.1 | - | 2 | 725 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY763517.1 | - | 2 | 202 | Chr. 18 - 50013635 - 50016315 [-] on Build GRCm38 |
