siRNA Details
| Gene Symbol: 2900072D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013764.1 | - | 1 | 896 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV154918.1 | - | 1 | 195 | Chr. 4 - 96030461 - 96030744 [+] on Build GRCm38 | |

2900072D07Rik
Genomic Map