siRNA Details
| Gene Symbol: 2900072G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013766.1 | - | 1 | 1524 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV068989.1 | - | 481 | Chr. - - [-] on Build | ||

2900072G11Rik
Genomic Map