siRNA Details
Gene Symbol: 4930488N15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015648.1 | - | 3 | 785 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715206.1 | - | 3 | 785 | Chr. 8 - 115585392 - 115594567 [+] on Build GRCm38 |
