siRNA Details
| Gene Symbol: Lrrc75aos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019771.1 | - | 4 | 615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc75aos1| Gene Symbol: Lrrc75aos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019771.1 | - | 4 | 615 | ||