siRNA Details
| Gene Symbol: Dscam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016518.1 | - | 5 | 2233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dscam| Gene Symbol: Dscam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016518.1 | - | 5 | 2233 | ||