siRNA Details
| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045976.1 | 3 | 520 | |||
| GenBank mRNA | AK016040.1 | - | 3 | 521 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715626.1 | - | 3 | 521 | Chr. 13 - 114607228 - 114689910 [+] on Build GRCm38 | |

4930544M13Rik
Genomic Map