siRNA Details
| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045976.1 | 3 | 603 | |||
| GenBank mRNA | AK016040.1 | - | 3 | 605 | ||
| AK076988.1 | - | 3 | 635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930544M13Rik| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045976.1 | 3 | 603 | |||
| GenBank mRNA | AK016040.1 | - | 3 | 605 | ||
| AK076988.1 | - | 3 | 635 | |||