siRNA Details
| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045976.1 | 5, 6 | 870 | |||
| GenBank mRNA | AK016040.1 | - | 5, 6 | 872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930544M13Rik| Gene Symbol: 4930544M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045976.1 | 5, 6 | 870 | |||
| GenBank mRNA | AK016040.1 | - | 5, 6 | 872 | ||