siRNA Details
| Gene Symbol: 4931440J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045503.1 | 5 | 396 | |||
| GenBank mRNA | AK016511.1 | - | 5 | 397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716190.1 | - | 5 | 394 | Chr. 14 - 62760503 - 62830127 [+] on Build GRCm38 | |

4931440J10Rik
Genomic Map