siRNA Details
| Gene Symbol: 4931440J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045503.1 | 7 | 838 | |||
| GenBank mRNA | AK016511.1 | - | 7 | 839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931440J10Rik| Gene Symbol: 4931440J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045503.1 | 7 | 838 | |||
| GenBank mRNA | AK016511.1 | - | 7 | 839 | ||