siRNA Details
| Gene Symbol: 4930545H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045357.1 | 2 | 270 | |||
| GenBank mRNA | AK016046.1 | - | 2 | 271 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715634.1 | - | 2 | 267 | Chr. 10 - 68319079 - 68321139 [-] on Build GRCm38 | |

4930545H06Rik
Genomic Map