siRNA Details
| Gene Symbol: 4930545E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045374.1 | 3 | 483 | |||
| GenBank mRNA | AK016045.1 | - | 3 | 484 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715633.1 | - | 3 | 484 | Chr. 18 - 17568782 - 17580990 [-] on Build GRCm38 | |

4930545E07Rik
Genomic Map