siRNA Details
| Gene Symbol: C330022C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045717.1 | 1 | 60 | |||
| GenBank mRNA | AK021219.1 | - | 1 | 61 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728818.1 | - | 1 | 61 | Chr. 7 - 140841193 - 140845689 [-] on Build GRCm38 | |

C330022C24Rik
Genomic Map