siRNA Details
Gene Symbol: C330022C24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045717.1 | 1 | 272 | |||
GenBank mRNA | AK021219.1 | - | 1 | 273 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728818.1 | - | 1 | 273 | Chr. 7 - 140841193 - 140845689 [-] on Build GRCm38 |
