siRNA Details
| Gene Symbol: C330022C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045717.1 | 4 | 1047 | |||
| GenBank mRNA | AK021219.1 | - | 4 | 1051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330022C24Rik| Gene Symbol: C330022C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045717.1 | 4 | 1047 | |||
| GenBank mRNA | AK021219.1 | - | 4 | 1051 | ||