siRNA Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021217.1 | - | 1 | 809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip1| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021217.1 | - | 1 | 809 | ||