siRNA Details
| Gene Symbol: 2900006G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013492.1 | - | 1 | 326 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712459.1 | - | 1 | 326 | Chr. 16 - 41222160 - 41222632 [+] on Build GRCm38 | |

2900006G07Rik
Genomic Map